missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Fractalkine / CX3CL1 Protein
A DNA sequence encoding the Mouse Fractalkine / CX3CL1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10262-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Specifications
NP_033168.2 | |
Fractalkine / CX3CL1 Protein | |
Research Use Only | |
CX3CL1 | |
The ED50 as determined by a cell proliferation assay using human peripheral blood lymphocytes (PBL) is less than 0.5 ug/ml, corresponding to a specific activity of > 2000 IU/mg. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20312 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
QHLGMTKCEI MCGKMTSRIP VALLIRYQLN QESCGKRAIV LETTQHRRFC ADPKEKWVQD AMKHLDHQAA ALTKNG | |
Greater than 97% as determined by SDS-PAGE and HPLC. |