missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Ephrin B3 / EFNB3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Ephrin B3 / EFNB3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5967-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_031937.1 | |
Ephrin B3 / EFNB3 Protein | |
Research Use Only | |
Efnb3 | |
Recombinant Protein | |
Yeast | |
LSLEPVYWNS ANKRFQAEGG YVLYPQIGDR LDLLCPRARP PGPHSSPSYE FYKLYLVEGA QGRRCEAPPA PNLLLTCDRP DLDLRFTIKF QEYSPNLWGH EFRSHHDYYI IATSDGTREG LESLQGGVCL TRGMKVLLRV GQSPRGGAVP RKPVSEMPME RDRGAAHSAE PGRDTIPGDP SSNATSRGAE GPLPPPSMPA | |
Greater than 90% as determined by SDS-PAGE. |
13643 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |