missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL14 / BRAK Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL14 / BRAK, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8658-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_062514.2 | |
CXCL14 / BRAK Protein | |
Research Use Only | |
Cxcl14 | |
Recombinant Protein | |
E. coli | |
SKCKCSRKGP KIRYSDVKKL EMKPKYPHCE EKMVIVTTKS MSRYRGQEHC LHPKLQSTKR FIKWYNAWNE KRRVYEE | |
Greater than 90% as determined by SDS-PAGE. |
57266 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |