missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL12 / SDF-1 Protein
A DNA sequence encoding the Mouse CXCL12 / SDF-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10258-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_038683.1 | |
CXCL12 / SDF-1 Protein | |
Research Use Only | |
CXCL12 | |
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 50-100 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20315 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK | |
Greater than 97% as determined by SDS-PAGE and HPLC. |