missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL1 / GRO-alpha Protein
Recombinant Protein
Marca: enQuireBio™ QP1455-1 mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_032202.1 | |
CXCL1 / GRO-alpha Protein | |
Research Use Only | |
CXCL1 | |
Recombinant Protein | |
E. coli | |
APIANELRCQ CLQTMAGIHL KNIQSLKVLP SGPHCTQTEV IATLKNGREA CLDPEAPLVQ KIVQKMLKGV PK | |
Greater than 95% as determined by SDS-PAGE. |
14825 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse CXCL1 / GRO-alpha induces Human neutrophil chemotaxis (significanty different from control at Mouse CXCL1 / GRO-alpha concentrations greater than 10 ng/ml). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |