missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CTGF Protein
A DNA sequence encoding the Mus musculus (Mouse) CTGF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5885-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_034347.2 | |
CTGF Protein | |
Research Use Only | |
CTGF | |
Recombinant Protein | |
E. coli | |
QDCSAQCQCA AEAAPHCPAG VSLVLDGCGC CRVCAKQLGE LCTERDPCDP HKGLFCDFGS PANRKIGVCT AKDGAPCVFG GSVYRSGESF QSSCKYQCTC LDGAVGCVPL CSMDVRLPSP DCPFPRRVKL PGKCCEEWVC DEPKDRTAVG PALAAYRLED TFGPDPTMMR ANCLVQTTEW SACSKTCGMG ISTRVTNDNT FCRLEKQSRL CMVRPCEADL EENIKKGKKC IRTPKIAKPV KFELSGCTSV KTYRAKFCGV CTDGRCCTPH RTTTLPVEFK CPDGEIMKKN MMFIKTCACH YNCPGDNDIF ESLYYRKMY | |
Greater than 80% as determined by SDS-PAGE. |
14219 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |