missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Complement C3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Complement C3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7399-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_033908.2 | |
Complement C3 Protein | |
Research Use Only | |
C3 | |
Recombinant Protein | |
E. coli | |
SEETKQNEAF SLTAKGKGRG TLSVVAVYHA KLKSKVTCKK FDLRVSIRPA PETAKKPEEA KNTMFLEICT KYLGDVDATM SILDISMMTG FAPDTKDLEL LASGVDRYIS KYEMNKAFSN KNTLIIYLEK ISHTEEDCLT FKVHQYFNVG LIQPGSVKVY SYYNLEESCT RFYHPEKDDG MLSKLCHSEM CRCAEENCFM QQSQEKINLN VRLDKACEPG VDYVYKTELT NIELLDDFDE YTMTIQQVIK SGSDEVQAGQ QRKFISHIKC RNALKLQKGK KYLMWGLSSD LWGEKPNTSY IIGKDTWVEH WPEAEECQDQ KYQKQCEELG AFTESMVVYG CPN | |
Greater than 80% as determined by SDS-PAGE. |
12266 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |