missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD82 / KAI-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CD82 / KAI-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5799-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001129527.1 | |
CD82 / KAI-1 Protein | |
Research Use Only | |
Cd82 | |
Recombinant Protein | |
E. coli | |
DKLKKEMGNT VMDIIRNYTA NATSSREEAW DYVQAQVKCC GWVSHYNWTE NEELMGFTKT TYPCSCEKIK EEDNQLIVKK GFCEADNSTV SENNPEDWPV NTEGCMEKAQ AWLQENF | |
Greater than 90% as determined by SDS-PAGE. |
12521 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |