missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD46 Protein
A DNA sequence encoding the Mus musculus (Mouse) CD46, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5790-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_034908.1 | |
CD46 Protein | |
Research Use Only | |
Cd46 | |
Recombinant Protein | |
E. coli | |
CELPRPFEAM ELKGTPKLFY AVGEKIEYKC KKGYLYLSPY LMIATCEPNH TWVPISDAGC IKVQCTMLQD PSFGKVYYID GSFSWGARAK FTCMEGYYVV GMSVLHCVLK GDDEAYWNGY PPHCEKIYCL PPPKIKNGTH TLTDINVFKY HEAVSYSCDP TPGPDKFSLV GTSMIFCAGH NTWSNSPPEC KVVKCPNPVL QNGRLISGAG EIFSYQSTVM FECLQGFYME GSSMVICSAN NSWEPSIPKC LKGPRPTHPT KPPVYNYTGY PSPREGIFSQ ELDAW | |
Greater than 90% as determined by SDS-PAGE. |
17221 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |