missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD122 / IL-2RB Protein
A DNA sequence encoding the Mus musculus (Mouse) CD122 / IL-2RB, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6219-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_032394.1 | |
CD122 / IL-2RB Protein | |
Research Use Only | |
IL2RB | |
Recombinant Protein | |
Yeast | |
AVKNCSHLEC FYNSRANVSC MWSHEEALNV TTCHVHAKSN LRHWNKTCEL TLVRQASWAC NLILGSFPES QSLTSVDLLD INVVCWEEKG WRRVKTCDFH PFDNLRLVAP HSLQVLHIDT QRCNISWKVS QVSHYIEPYL EFEARRRLLG HSWEDASVLS LKQRQQWLFL EMLIPSTSYE VQVRVKAQRN NTGTWSPWSQ PLTFRTRPAD PMKE | |
Greater than 90% as determined by SDS-PAGE. |
16185 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |