missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL5 / RANTES Protein
A DNA sequence encoding the Mouse CCL5 / RANTES, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10265-ec-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_038681.2 | |
CCL5 / RANTES Protein | |
Research Use Only | |
CCL5 | |
The biological activity determined by a chemotaxis bioassay using human T lymphocytes is in a concentration range of 1.0-10 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20304 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS | |
Greater than 97% as determined by SDS-PAGE and HPLC. |