missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL28 Protein
A DNA sequence encoding the Mouse CCL28, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10250-ec-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_064675.1 | |
CCL28 Protein | |
Research Use Only | |
Ccl28 | |
The biological activity determined by a chemotaxis bioassay using murine lymphocytes is in a concentration range of 1.0-10.0 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
56838 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SEAILPMASS CCTEVSHHVS GRLLERVSSC SIQRADGDCD LAAVILHVKR RRICISPHNR TLKQWMRASE VKKNGRENVC SGKKQPSRKD RKGHTTRKHR TRGTHRHEAS R | |
Greater than 97% as determined by SDS-PAGE and HPLC. |