missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C10ORF54 Protein
A cDNA sequence encoding the C10ORF54 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11219-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
C10ORF54 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
FKVTTPYSLY VCPEGQNATL TCRILGPVSK GHDVTIYKTW YLSSRGEVQM CKEHRPIRNF TLQHLQHHGS HLKANASHDQ PQKHGLELAS DHHGNFSITL RNVTPRDSGL YCCLVIELKN HHPEQRFYGS MELQVQAGKG SGSTCMASNE QDSDSITAAL EHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
C10ORF54 protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |