missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C-C motif chemokine 21c Protein
A DNA sequence encoding the Mouse C-C motif chemokine 21c, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10275-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001180595.1 | |
C-C motif chemokine 21c Protein | |
Research Use Only | |
Ccl21c | |
The biological activity determined by a chemotaxis bioassay using murine T-lymphocytes is in a concentration range of 10-100 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
100041504 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SDGGGQDCCL KYSQKKIPYS IVRGYRKQEP SLGCPIPAIL FLPRKHSKPE LCANPEEGWV QNLMRRLDQP PAPGKQSPGC RKNRGTSKSG KKGKGSKGCK RTEQTQPSRG | |
Greater than 97% as determined by SDS-PAGE and HPLC. |