missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Annexin A1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Annexin A1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5650-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_034860.2 | |
Annexin A1 Protein | |
Research Use Only | |
Anxa1 | |
Recombinant Protein | |
Yeast | |
AMVSEFLKQA RFLENQEQEY VQAVKSYKGG PGSAVSPYPS FNVSSDVAAL HKAIMVKGVD EATIIDILTK RTNAQRQQIK AAYLQENGKP LDEVLRKALT GHLEEVVLAM LKTPAQFDAD ELRGAMKGLG TDEDTLIEIL TTRSNEQIRE INRVYREELK RDLAKDITSD TSGDFRKALL ALAKGDRCQD LSVNQDLADT DARALYEAGE RRKGTDVNVF TTILTSRSFP HLRRVFQNYG KYSQHDMNKA LDLELKGDIE KCLTTIVKCA TSTPAFFAEK LYEAMKGAGT RHKALIRIMV SRSEIDMNEI KVFYQKKYGI SLCQAILDET KGDYEKILVA LCGGN | |
Greater than 90% as determined by SDS-PAGE. |
16952 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |