Learn More
enQuireBio™ Recombinant Mouse Angiogenin-4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Angiogenin-4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9784-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_808212.2 | |
Angiogenin-4 Protein | |
Research Use Only | |
Ang4 | |
Recombinant Protein | |
Yeast | |
QNERYEKFLR QHYDAKPNGR DDRYCESMMK ERKLTSPCKD VNTFIHGTKK NIRAICGKKG SPYGENFRIS NSPFQITTCT HSGASPRPPC GYRAFKDFRY IVIACEDGWP VHFDESFISP | |
Greater than 90% as determined by SDS-PAGE. |
219033 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |