missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse AK2 Protein
A cDNA sequence encoding the AK2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10510-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
AK2 Protein | |
Research Use Only | |
Specific activity is > 40 units/mg. One unit will convert 2.0 umoles of ADP to ATP + AMP per minute at pH 7.5 at 37C. | |
Mouse | |
His | |
AK2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.5), 10% glycerol and 1mM DTT. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAPNVL ASEPEIPKGI RAVLLGPPGA GKGTQAPKLA ENFCVCHLAT GDMLRAMVAS GSEL TMDAGKLVSD EMVVELIEKN LETPSCKNGF LLDGFPRTVR QAEMLDDLME KRKEKLDSVI EFSIQDSLLI RRITGRLIHP KSGRSNPPKEPMKDD ITGEPLIRRS DDNEKALKTR LEAYHTQTTP LVEYYRKRGI HCAIDASQTP DIVFASILAA FSKATCKDLV MFI | |
Greater than 95.0% as determined by SDS-PAGE. |