missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Agxt, mitochondrial Protein
A DNA sequence encoding the Mus musculus (Mouse) Agxt, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5628-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_057911.2 | |
Agxt, mitochondrial Protein | |
Research Use Only | |
Agxt | |
Recombinant Protein | |
E. coli | |
SYQLLVPPPE ALSKPLSVPT RLLLGPGPSN LAPRVLAAGS LRMIGHMQKE MLQIMEEIKQ GIQYVFQTRN PLTLVVSGSG HCAMETALFN LLEPGDSFLT GTNGIWGMRA AEIADRIGAR VHQMIKKPGE HYTLQEVEEG LAQHKPVLLF LVHGESSTGV VQPLDGFGEL CHRYQCLLLV DSVASLGGVP IYMDQQGIDI MYSSSQKVLN APPGISLISF NDKAKYKVYS RKTKPVSFYT DITYLAKLWG CEGETRVIHH TTPVTSLYCL RESLALIAEQ GLENCWRRHR EATAHLHKHL QEMGLKFFVK DPEIRLPTIT TVTVPAGYNW RDIVSYVLDH FSIEISGGLG PTEERVLRIG LLGYNATTEN VDRVAEALRE ALQHCPKNKL | |
Greater than 90% as determined by SDS-PAGE. |
11611 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |