missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Acid ceramidase Protein
A DNA sequence encoding the Mus musculus (Mouse) Acid ceramidase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8262-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_062708.1 | |
Acid ceramidase Protein | |
Research Use Only | |
ASAH1 | |
Recombinant Protein | |
E. coli | |
QAQDVPPWTE DCRKSTYPPS GPTYRGPVPW HTINLDLPPY KRWHELLAQK APALRILVNS ITSLVNTFVP SGKLMKMVDQ KLPGMIGSLP DPFGEEMRGI ADVTGIPLGE IISFNIFYEL FTM | |
Greater than 80% as determined by SDS-PAGE. |
11886 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-GST | |
Tris-based buffer, 50% glycerol |