missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque Serum Albumin / HSA / ALB Protein
A DNA sequence encoding the Macaca fascicularis (Crab-eating macaque) (Cynomolgus monkey) Serum Albumin / HSA / ALB, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5643-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001306284.1 | |
Serum Albumin / HSA / ALB Protein | |
Research Use Only | |
ALB | |
Recombinant Protein | |
E. coli | |
DTHKSEVAHR FKDLGEEHFK GLVLVAFSQY LQQCPFEEHV KLVNEVTEFA KTCVADESAE NCDKSLHTLF GDKLCTVATL RETYGEMADC CAKQEPERNE CFLQHKDDNP NLPPLVRPEV DVMCTAFHDN EATFLKKYLY EVARRHPYFY APELLFFAAR YKAAFAECCQ AADKAACLLP KLDELRDEGK ASSAKQRLKC ASLQKFGDRA FKAWAVARLS QKFPKAEFAE VSKLVTDLTK VHTECCHGDL LECADDRADL AKYMCENQDS ISSKLKECCD KPLLEKSHCL AEVENDEMPA DLPSLAADYV ESKDVCKNYA EAKDVFLGMF LYEYARRHPD YSVMLLLRLA KAYEATLEKC CAAADPHECY AKVFDEFQPL VEEPQNLVKQ NCELFEQLGE YKFQNALLVR YTKKVPQVST PTLVEVSRNL GKVGAKCCKL PEAKRMPCAE DYLSVVLNRL CVLHEKTPVS EKVTKCCTES LVNRRPCFSA LELDEAYVPK AFNAETFTFH ADMCTLSEKE KQVKKQTALV ELVKHKPKAT KEQLKGVMDN FAAFVEKCCK ADDKEACFAE EGPKFVAASQ AALA | |
Greater than 90% as determined by SDS-PAGE. |
102130757 | |
50 μg | |
Not determined. | |
Full Length | |
Macaque | |
His-SUMO | |
Tris-based buffer, 50% glycerol |