missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Shandong Protein
Recombinant Protein
Marca: enQuireBio™ QP12086-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Influenza H3N2 Shandong Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The H3N2A/ Shandong/9/93 solution (1.0 mg/ml) contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE. |