missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zymogen granule protein 16 homolog B Protein
A DNA sequence encoding the Homo sapiens (Human) Zymogen granule protein 16 homolog B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7869-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_660295.2 | |
Zymogen granule protein 16 homolog B Protein | |
Research Use Only | |
ZG16B | |
Recombinant Protein | |
E. coli | |
GKMYGPGGGK YFSTTEDYDH EITGLRVSVG LLLVKSVQVK LGDSWDVKLG ALGGNTQEVT LQPGEYITKV FVAFQAFLRG MVMYTSKDRY FYFGKLDGQI SSAYPSQEGQ VLVGIYGQYQ LLGIKSIGFE WNYPLEEPTT EPPVNLTYSA NSPVGR | |
Greater than 90% as determined by SDS-PAGE. |
124220 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |