missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WNT7A Protein
A cDNA sequence encoding the WNT7A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13963-20ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
7476 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLGASIIC NKIPGLAPRQ RAICQSRPDA IIVIGEGSQM GLDECQFQFR NGRWNCSALG ERTVFGKELK VGSREAAFTY AIIAAGVAHA ITAACTQGNL SDCGCDKEKQ GQYHRDEGWK WGGCSADIRY GIGFAKVFVD AREIKQNART LMNLHNNEAG RKILEENMKL ECKCHGVSGS CTTKTCWTTL PQFRELGYVL KDKYNEAVHV EPVRASRNKR PTFLKIKKPL SYRKPMDTDL VYIEKSPNYC EEDPVTGSVG TQGRACNKTA PQASGCDLMC CGRGYNTHQY ARVWQCNCKF HWCCYVKCNT CSERTEMYTC K | |
Greater than 85.0% as determined by SDS-PAGE. |
WNT7A Protein | |
Research Use Only | |
WNT7A | |
Human | |
His | |
WNT7A protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |