missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VPS13A Protein
A DNA sequence encoding the Homo sapiens (Human) VPS13A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7966-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001018047.1 | |
VPS13A Protein | |
Research Use Only | |
VPS13A | |
Recombinant Protein | |
E. coli | |
RPPRFFNEDG VIRPYRLRDG TGNQMLQVME NGRFAKYKYF THVMINKTDM LMITRRGVLF VTKGTFGQLT CEWQYSFDEF TKEPFIVHGR RLRIEAKERV KSVF | |
Greater than 90% as determined by SDS-PAGE. |
23230 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |