missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VAMP5 Protein
A cDNA sequence encoding the VAMP5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13919-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
VAMP5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMAG IELERCQQQA NEVTEIMRNN FGKVLERGVK LAELQQRSDQ LLDMSSTFNK TTQNLAQKKC WENIRYRIC | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The VAMP5 protein solution contains 20mM Tris-HCl, pH 8.0, 100mM NaCl, 1mM DTT and 10% glycerol. |