missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UFSP1 Protein
A cDNA sequence encoding the UFSP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13883-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
UFSP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGDKPPG FRGSRDWIGC VEASLCLAHF GGPQGRLCHV PRGVGLHGEL ERLYSHFAGG GGPVMVGGDA DARSKALLGV CVGSGTEAYV LVLDPHYWGT PKSPSELQAA GWVGWQEVSA AFDPNSFYNL CLTSLSSQQQ QRTLD | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UFSP1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.5), 0.2M NaCl, 30% glycerol and 1mM DTT. |