missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UCHL1 / PGP9.5 Protein
A DNA sequence encoding the Homo sapiens (Human) UCHL1 / PGP9.5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8722-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Spezifikation
NP_004172.2 | |
UCHL1 / PGP9.5 Protein | |
Research Use Only | |
UCHL1 | |
Recombinant Protein | |
E. coli | |
MQLKPMEINP EMLNKVLSRL GVAGQWRFVD VLGLEEESLG SVPAPACALL LLFPLTAQHE NFRKKQIEEL KGQEVSPKVY FMKQTIGNSC GTIGLIHAVA NNQDKLGFED GSVLKQFLSE TEKMSPEDRA KCFEKNEAIQ AAHDAVAQEG QCRVDDKVNF HFILFNNVDG HLYELDGRMP FPVNHGASSE DTLLKDAAKV CREFTEREQG EVRFSAVALC KA | |
Greater than 90% as determined by SDS-PAGE. |
7345 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |