missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBXN2B Protein
A cDNA sequence encoding the UBXN2B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13878-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
UBXN2B Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UBXN2B solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEGGGP EPGEQERRSS GPRPPSARDL QLALAELYED EVKCKSSKSN RPKATVFKSP RTPPQRFYSS EHEYSGLNIV RPSTGKIVNE LFKEAREHGA VPLNEATRAS GDDKSKSFTG GGYRLGSSFC KRSEYIYGEN QLQDVQILLK LWSNGFSLDD GELRPYNEPT NAQFLESVKR GEIPLELQRL VHGGQVNLDM EDHQDQEYIK PRLRFKAFSG EGQKLGSLTP EIVSTPSSPE EEDKSILNAV VLIDDSVPTT KIQIRLADGS RLIQRFNSTH RILDVRNFIV QSRPEFAALD FILVTSFPNK ELTDESLTLL EADILNTVLL QQLK |