missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBL3 Protein
A cDNA sequence encoding the UBL3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13872-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
UBL3 Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UBL3 protein solution (1 mg/1ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 2mM DTT. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSNVPADMI NLRLILVSGK TKEFLFSPND SASDIAKHVY DNWPMDWEEE QVSSPNILRL IYQGRFLHGN VTLGALKLPF GKTTVMHLVA RETLPEPNSQ GQRNREKTGE SNCC |