missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin-like protein ISG15 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-like protein ISG15, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8673-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_056598.2 | |
Ubiquitin-like protein ISG15 Protein | |
Research Use Only | |
G1P2 | |
Recombinant Protein | |
E. coli | |
AWDLKVKMLG GNDFLVSVTN SMTVSELKKQ IAQKIGVPAF QQRLAHQTAV LQDGLTLSSL GLGPSSTVML VVQNCSEPLS ILVRNERGHS NIYEVFLTQT VDTLKKKVSQ REQVHEDQFW LSFEGRPMED KELLGEYGLK PQCTVIKHLR LRGGGG | |
Greater than 90% as determined by SDS-PAGE. |
100038882 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |