missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 K Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-conjugating enzyme E2 K, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8668-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001104582.1 | |
Ubiquitin-conjugating enzyme E2 K Protein | |
Research Use Only | |
UBE2K | |
Recombinant Protein | |
E. coli | |
ANIAVQRIKR EFKEVLKSEE TSKNQIKVDL VDENFTELRG EIAGPPDTPY EGGRYQLEIK IPETYPFNPP KVRFITKIWH PNISSVTGAI CLDILKDQWA AAMTLRTVLL SLQALLAAAE PDDPQDAVVA NQYKQNPEMF KQTARLWAHV YAGAPVSSPE YTKKIENLCA MGFDRNAVIV ALSSKSWDVE TATELLLSN | |
Greater than 90% as determined by SDS-PAGE. |
3093 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |