missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2F Protein
A DNA sequence encoding the Homo sapiens (Human) UBE2F, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7958-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001265234.1 | |
UBE2F Protein | |
Research Use Only | |
UBE2F | |
Recombinant Protein | |
E. coli | |
MLTLASKLKR DDGLKGSRTA ATASDSTRRV SVRDKLLVKE VAELEANLPC TCKVHFPDPN KLHCFQLTVT PDEGYYQGGK FQFETEVPDA YNMVPPKVKC LTKIWHPNIT ETGEICLSLL REHSIDGTGW APTRTLKDVV WGLNSLFTDL LNFDDPLNIE AAEHHLRDKE DFRNKVDDYI KRYAR | |
Greater than 80% as determined by SDS-PAGE. |
140739 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |