missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2E1 Protein
A cDNA sequence encoding the UBE2E1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13857-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
UBE2E1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSDDDSR ASTSSSSSSS SNQQTEKETN TPKKKESKVS MSKNSKLLST SAKRIQKELA DITLDPPPNC SAGPKGDNIY EWRSTILGPP GSVYEGGVFF LDITFTPEYP FKPPKVTFRT RIYHCNINSQ GVICLDILKD NWSPALTISK VLLSICSLLT DCNPADPLVG SIATQYMTNR AEHDRMARQW TKRYAT | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UBE2E1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 20% glycerol. |