missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2D2 Protein
A cDNA sequence encoding the UBE2D2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13856-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
UBE2D2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MALKRIHKEL NDLARDPPAQ CSAGPVGDDM FHWQATIMGP NDSPYQGGVF FLTIHFPTDY PFKPPKVAFT TRIYHPNINS NGSICLDILR SQWSPALTIS KVLLSICSLL CDPNPDDPLV PEIARIYKTD REKYNRIARE WTQKYAM | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The UBE2D2 solution (0.5 mg/ml) contains 20mM MES (pH6.0), 50mM NaCl and 1mM DTT. |