missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human U1 small nuclear ribonucleoprotein 1
A DNA sequence encoding the Homo sapiens (Human) U1 small nuclear ribonucleoprotein A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8341-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004587.1 | |
U1 small nuclear ribonucleoprotein 1 | |
Research Use Only | |
SNRPA | |
Recombinant Protein | |
E. coli | |
PNHTIYINNL NEKIKKDELK KSLYAIFSQF GQILDILVSR SLKMRGQAFV IFKEVSSATN ALRSMQGFPF YDKPMRIQYA KTDSDIIAKM KGTFVERDRK REKRKPKSQE TPATKKAVQG GGATPVVGAV QGPVPGMPPM TQAPRIMHHM PGQPPYMPPP GMIPPPGLAP GQIPPGAMPP QQLMPGQMPP AQPLSENPPN HILFLTNLPE ETNELMLSML FNQFPGFKEV RLVPGRHDIA FVEFDNEVQA GAARDALQGF KITQNNAMKI S | |
Greater than 90% as determined by SDS-PAGE. |
6626 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |