missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSSC4 Protein
A cDNA sequence encoding the TSSC4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13831-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TSSC4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAEAGT GEPSPSVEGE HGTEYDTLPS DTVSLSDSDS DLSLPGGAEV EALSPMGLPG EEDSGPDEPP SPPSGLLPAT VQPFHLRGMS STFSQRSRDI FDCLEGAARR APSSVAHTSM SDNGGFKRPL APSGRSPVEG LGRAHRSPAS PRVPPVPDYV AHPERWTKYS LEDVTEVSEQ SNQATALAFL GSQSLAAPTD CVSSFNQDPS SCGEGRVIFT KPVRGVEARH ERKRVLGKVG EPGRGGLGNP ATDRGEGPVE LAHLAGPGSP EAEEWGSPHG GLQEVEALSG SVHSGSVPGL PPVETVGFHG SRKRSRDHFR NKSSSPEDPG AEV | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TSSC4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 20% glycerol. |