missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSLP Protein
A DNA sequence encoding the Human TSLP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10350-ec-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_149024.1 | |
TSLP Protein | |
Research Use Only | |
TSLP | |
The ED50 as determined by a cell proliferation assay using human IL-7Ralpha and human TSLP R co-transfected murine BaF3 pro-B cells is less than 0.3 ng/ml, corresponding to a specific activity of > 3.3 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150 mM NaCl, lyophilized |
85480 | |
2 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MYDFTNCDFE KIKAAYLSTI SKDLITYMSG TKSTEFNNTV SCSNRPHCLT EIQSLTFNPT AGCASLAKEM FAMKTKAALA IWCPGYSETQ INATQAMKKR RKRKVTTNKC LEQVSQLQGL WRRFNRPLLK QQ | |
Greater than 98% as determined by SDS-PAGE and HPLC. |