missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trypsin-3 / PRSS3 Protein
A DNA sequence encoding the Homo sapiens (Human) Trypsin-3 / PRSS3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8742-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001184026.2 | |
Trypsin-3 / PRSS3 Protein | |
Research Use Only | |
PRSS3 | |
Recombinant Protein | |
E. coli | |
IVGGYTCEEN SLPYQVSLNS GSHFCGGSLI SEQWVVSAAH CYKTRIQVRL GEHNIKVLEG NEQFINAAKI IRHPKYNRDT LDNDIMLIKL SSPAVINARV STISLPTTPP AAGTECLISG WGNTLSFGAD YPDELKCLDA PVLTQAECKA SYPGKITNSM FCVGFLEGGK DSCQRDSGGP VVCNGQLQGV VSWGHGCAWK NRPGVYTKVY NYVDWIKDTI AAN | |
Greater than 80% as determined by SDS-PAGE. |
5646 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |