missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trypsin-1 Protein
A cDNA sequence encoding the Trypsin-1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10898-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Trypsin-1 Protein | |
Research Use Only | |
1 unit hydrolyzes 1 micromole of Na-Benzoyl-D,L-arginine 4-nitroanilide (BANI) per minute at 37C, pH 8.0, in the presence of 10mM calcium. The specific activity is >1,000 Units per mg protein as measured under the described conditions. | |
Human | |
Untagged | |
The Trypsin-1 filtered 0.22um solution contains 20mM TRIS-HCl buffer pH 7.2. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human | |
IVGGYNCEEN SVPYQVSLNS GYHFCGGSLI NEQWVVSAGH CYKSRIQVRL GEHNIEVLEG NEQFINAAKI IRHPQYDRKT LNNDIMLIKL SSRAVINARV STISLPTAPP ATGTKCLISG WGNTASSGAD YPDELQCLDA PVLSQAKCEA SYPGKITSNM FCVGFLEGGK DSCQGDSGGP VVCNGQLQGV VSWGDGCAQK NKPGVYTKVY NYVKWIKNTI AANS | |
Greater than 95% as determined by SDS-PAGE. |