missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRPA1 Protein
A DNA sequence encoding the Homo sapiens (Human) TRPA1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6843-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_015628.2 | |
TRPA1 Protein | |
Research Use Only | |
TRPA1 | |
Recombinant Protein | |
E. coli | |
IGLAVGDIAE VQKHASLKRI AMQVELHTSL EKKLPLWFLR KVDQKSTIVY PNKPRSGGML FHIFCFLFCT GEIRQEIPNA DKSLEMEILK QKYRLKDLTF LLEKQHELIK LIIQKMEIIS ETEDDDSHCS FQDRFKKEQM EQRNSRWNTV LRAVKAKTHH LEP | |
Greater than 90% as determined by SDS-PAGE. |
8989 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |