missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transmembrane protease serine 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Transmembrane protease serine 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9439-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001128571.1 | |
Transmembrane protease serine 2 Protein | |
Research Use Only | |
TMPRSS2 | |
Recombinant Protein | |
Yeast | |
WKFMGSKCSN SGIECDSSGT CINPSNWCDG VSHCPGGEDE NRCVRLYGPN FILQVYSSQR KSWHPVCQDD WNENYGRAAC RDMGYKNNFY SSQGIVDDSG STSFMKLNTS AGNVDIYKKL YHSDACSSKA VVSLRCIACG VNLNSSRQSR IVGGESALPG AWPWQVSLHV QNVHVCGGSI ITPEWIVTAA HCVEKPLNNP WHWTAFAGIL RQSFMFYGAG YQVEKVISHP NYDSKTKNND IALMKLQKPL TFNDLVKPVC LPNPGMMLQP EQLCWISGWG ATEEKGKTSE VLNAAKVLLI ETQRCNSRYV YDNLITPAMI CAGFLQGNVD SCQGDSGGPL VTSKNNIWWL IGDTSWGSGC AKAYRPGVYG NVMVFTDWIY RQMRADG | |
Greater than 90% as determined by SDS-PAGE. |
7113 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |