missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human tPA / PLAT Protein
A DNA sequence encoding the Homo sapiens (Human) tPA / PLAT, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6501-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000921.1 | |
tPA / PLAT Protein | |
Research Use Only | |
PLAT | |
Recombinant Protein | |
E. coli | |
SYQVICRDEK TQMIYQQHQS WLRPVLRSNR VEYCWCNSGR AQCHSVPVKS CSEPRCFNGG TCQQALYFSD FVCQCPEGFA GKCCEIDTRA TCYEDQGISY RGTWSTAESG AECTNWNSSA LAQKPYSGRR PDAIRLGLGN HNYCRNPDRD SKPWCYVFKA GKYSSEFCST PACSEGNSDC YFGNGSAYRG THSLTESGAS CLPWNSMILI GKVYTAQNPS AQALGLGKHN YCRNPDGDAK PWCHVLKNRR LTWEYCDVPS CSTCGLRQYS QPQFRIKGGL FADIASHPWQ AAIFAKHRRS PGERFLCGGI LISSCWILSA AHCFQERFPP HHLTVILGRT YRVVPGEEEQ KFEVEKYIVH KEFDDDTYDN DIALLQLKSD SSRCAQESSV VRTVCLPPAD LQLPDWTECE LSGYGKHEAL SPFYSERLKE AHVRLYPSSR CTSQHLLNRT VTDNMLCAGD TRSGGPQANL HDACQGDSGG PLVCLNDGRM TLVGIISWGL GCGQKDVPGV YTKVTNYLDW I | |
Greater than 90% as determined by SDS-PAGE. |
5327 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |