missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFSF12 Protein
A DNA sequence encoding the Homo sapiens (Human) TNFSF12, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6819-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_003800.1 | |
TNFSF12 Protein | |
Research Use Only | |
TNFSF12 | |
Recombinant Protein | |
E. coli | |
SLGSRASLSA QEPAQEELVA EEDQDPSELN PQTEESQDPA PFLNRLVRPR RSAPKGRKTR ARRAIAAHYE VHPRPGQDGA QAGVDGTVSG WEEARINSSS PLRYNRQ | |
Greater than 90% as determined by SDS-PAGE. |
407977 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |