missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFAIP6 Protein
A DNA sequence encoding the Homo sapiens (Human) TNFAIP6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6813-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_009046.2 | |
TNFAIP6 Protein | |
Research Use Only | |
TNFAIP6 | |
Recombinant Protein | |
E. coli | |
WGFKDGIFHN SIWLERAAGV YHREARSGKY KLTYAEAKAV CEFEGGHLAT YKQLEAARKI GFHVCAAGWM AKGRVGYPIV KPGPNCGFGK TGIIDYGIRL NRSERWDAYC YNPHAKECGG VFTDPKQIFK SPGFPNEYED NQICYWHIRL KYGQRIHLSF LDFDLEDDPG CLADYVEIYD SYDDVHGFVG RYCGDELPDD IISTGNVMTL KFLSDASVTA GGFQIKYVAM DPVSKSSQGK NTSTTSTGNK NFLAGRFSHL | |
Greater than 90% as determined by SDS-PAGE. |
7130 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |