missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TLR2 / CD282 Protein
A DNA sequence encoding the Homo sapiens (Human) TLR2 / CD282, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6800-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001305716.1 | |
TLR2 / CD282 Protein | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7097 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
TLR2 | |
Recombinant Protein | |
E. coli | |
KEESSNQASL SCDRNGICKG SSGSLNSIPS GLTEAVKSLD LSNNRITYIS NSDLQRCVNL QALVLTSNGI NTIEEDSFSS LGSLEHLDLS YNYLSNLSSS WFKPLSSLTF LNLLGNPYKT LGETSLFSHL TKLQILRVGN MDTFTKIQRK DFAGLTFLEE LEIDASDLQS YEPKSLKSIQ NVSHLILHMK QHILLLEIFV DVTSSVECLE LRDTDLDTFH FSELSTGETN SLIKKFTFRN VKITDESLFQ VMKLLNQISG LLELEFDDCT LNGVGNFRAS DNDRVIDPGK VETLTIRRLH IPRFYLFYDL STLYSLTERV KRITVENSKV FLVPCLLSQH LKSLEYLDLS ENLMVEEYLK NSACEDAWPS LQTLILRQNH LASLEKTGET LLTLKNLTNI DISKNSFHSM PETCQWPEKM KYLNLSSTRI HSVTGCIPKT LEILDVSNNN LNLFSLNLPQ LKELYISRNK LMTLPDASLL PMLLVLKISR NAITTFSKEQ LDSFHTLKTL EAGGNNFICS CEFLSFTQEQ QALAKVLIDW PANYLCDSPS HVRGQQVQDV RLSVSECHRT |