Learn More
enQuireBio™ Recombinant Human Thyroid hormone-inducible hepatic Protein
A DNA sequence encoding the Homo sapiens (Human) Thyroid hormone-inducible hepatic protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7931-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_003242.1 | |
Thyroid hormone-inducible hepatic Protein | |
Research Use Only | |
Thrsp | |
Recombinant Protein | |
E. coli | |
MQVLTKRYPK NCLLTVMDRY AAEVHNMEQV VMIPSLLRDV QLSGPGGQAQ AEAPDLYTYF TMLKAICVDV DHGLLPREEW QAKVAGSEEN GTAETEEVED ESASGELDLE AQFHLHFSSL HHILMHLTEK AQEVTRKYQE MTGQVW | |
Greater than 80% as determined by SDS-PAGE. |
7069 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |