missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thiamin pyrophosphokinase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Thiamin pyrophosphokinase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6827-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001035947.1 | |
Thiamin pyrophosphokinase 1 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
27010 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
TPK1 | |
Recombinant Protein | |
E. coli | |
MEHAFTPLEP LLSTGNLKYC LVILNQPLDN YFRHLWNKAL LRACADGGAN RLYDITEGER ESFLPEFING DFDSIRPEVR EYYATKGCEL ISTPDQDHTD FTKCLKMLQK KIEEKDLKVD VIVTLGGLAG RFDQIMASVN TLFQATHITP FPIIIIQEES LIYLLQPGKH RLHVDTGMEG DWCGLIPVGQ PCMQVTTTGL KWNLTNDVLA FGTLVSTSNT YDGSGVVTVE TDHPLLWTMA IKS |