missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human THG1L Protein
A cDNA sequence encoding the THG1L was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13742-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
54974 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAKSKFE YVRDFEADDT CLAHCWVVVR LDGRNFHRFA EKHNFAKPND SRALQLMTKC AQTVMEELED IVIAYGQSDE YSFVFKRKTN WFKRRASKFM THVASQFASS YVFYWRDYFE DQPLLYPPGF DGRVVVYPSN QTLKDYLSWR QADCHINNLY NTVFWALIQQ SGLTPVQAQG RLQGTLAADK NEILFSEFNI NYNNELPMYR KGTVLIWQKV DEVMTKEIKL PTEMEGKKMA VTRTRTKPVP LHCDIIGDAF WKEHPEILDE DS | |
Greater than 90% as determined by SDS-PAGE. |
THG1L Protein | |
Research Use Only | |
THG1L | |
Human | |
His | |
The THG1L solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol. |