missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFBR3 Protein
A cDNA sequence encoding the TGFBR3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10501-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TGFBR3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
PYSNPDRMSH YTIIENICPK DESVKFYSPK RVHFPIPQAD MDKKRFSFVF KPVFNTSLLF LQCELTLCTK MEKHPQKLPK CVPPDEACTS LDASIIWAMM QNKKTFTKPL AVIHHEAESK EKGPSMKEPN PISPPIFHGL DTLTVMGIAF AAFVIGALLT GALWYIYSHT GETAGRQQVP TSPPASENSS AAHSIGSTQS TPCSSSS |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
GST | |
100microL purified human TGF Beta Receptor 3 protein at 100microg/ml in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |