missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TFRC Protein
A cDNA sequence encoding the TFRC was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13731-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TFRC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMCKGVEP KTECERLAGT ESPVREEPGE DFPAARRLYW DDLKRKLSEK LDSTDFTGTI KLLNENSYVP REAGSQKDEN LALYVENQFR EFKLSKVWRD QHFVKIQVKD SAQNSVIIVD KNGRLVYLVE NPGGYVAYSK AATVTGKLVH ANFGTKKDFE DLYTPVNGSI VIVRAGKITF AEKVANAESL NAIGVLIYMD QTKFPIVNAE LSFFGHAHLG TGDPYTPGFP SFNHTQFPPS RSSGLPNIPV QTISRAAAEK LFGNMEGDCP SDWKTDSTCR MVTSESKNVK LTVSNVLKEI KILNIFGVIK GFVEPDHYVV VGAQRDAWGP GAAKSGVGTA LLLKLAQMFS DMVLKDGFQP SRSIIFASWS AGDFGSVGAT EWLEGYLSSL HLKAFTYINL DKAVLGTSNF KVSASPLLYT LIEKTMQNVK HPVTGQFLYQ DSNWASKVEK LTLDNAAFPF LAYSGIPAVS FCFCEDTDYP YLGTTMDTYK ELIERIPELN KVARAAAEVA GQFVIKLTHD VELNLDYERY NSQLLSFVRD LNQYRADIKE MGLSLQWLYS ARGDFFRATS RLTTDFGNAE KTDRFVMKKL NDRVMRVEYH FLSPYVSPKE SPFRHVFWGS GSHTLPALLE NLKLRKQNNG AFNETLFRNQ LALATWTIQG AANALSGDVW DIDNEF | |
Greater than 80.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TFRC protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |